accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OCH3CH3HOClClClCH3CH3HOClClCl

CAS RN 6001-64-5

IUPAC Name: bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate
Synonyms: bis(chlorobutanol) hydrate
Molecular Weight (g/mol): 372.91
Molecular Formula: C8H16Cl6O3
InChi Key: WRWLCXJYIMRJIN-UHFFFAOYSA-N
SMILES: O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (8)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)
  • (6)
Boiling Point 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (6)
  • (1)
  • (3)
  • (6)
  • (2)
  • (5)
  • (6)
  • (3)
  • (3)
  • (4)
  • (6)
  • (2)
  • (1)
  • (8)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (6)