accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 6004-60-0

IUPAC Name: 1-cyclopentylethan-1-one
Synonyms: ethanone, 1-cyclopentyl-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: LKENTYLPIUIMFG-UHFFFAOYSA-N
SMILES: CC(=O)C1CCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)