accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)HO(R)OH(S)OOHO(S)HO

CAS RN 60046-25-5

IUPAC Name: (4S,5S)-3,4-dihydroxy-5-[(1R,2R)-1,2,3-trihydroxypropyl]oxolan-2-one
Synonyms: glucoheptonolactone
Molecular Weight (g/mol): 208.17
Molecular Formula: C7H12O7
InChi Key: VIVCRCODGMFTFY-JPRIQSOUSA-N
SMILES: OC[C@@H](O)[C@@H](O)[C@@H]1OC(=O)C(O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)