accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60047-17-8

IUPAC Name: 2-(5-ethenyl-5-methyloxolan-2-yl)propan-2-ol
Synonyms: linalool oxide
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: BRHDDEIRQPDPMG-UHFFFAOYNA-N
SMILES: CC(C)(O)C1CCC(C)(O1)C=C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results