accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6006-65-1

IUPAC Name: (dipropoxymethyl)dimethylazanium
Synonyms: (dipropoxymethyl)dimethylazanium
Molecular Weight (g/mol): 176.28
Molecular Formula: C9H22NO2
InChi Key: NSLGQFIDCADTAS-UHFFFAOYSA-O
SMILES: CCCOC(OCCC)[NH+](C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)