accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6006-95-7

H3C

CAS RN 6006-95-7

IUPAC Name: pentadecylcyclohexane
Synonyms: N-pentadecylcyclohexane
Molecular Weight (g/mol): 294.57
Molecular Formula: C21H42
InChi Key: ZNUABQHWFGTOCO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)