accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6007-83-6

IUPAC Name: 4H,6H-thieno[2,3-c]furan-4,6-dione
Synonyms: thieno[2,3-c]furan-4,6-dione
Molecular Weight (g/mol): 154.14
Molecular Formula: C6H2O3S
InChi Key: FIDKFEIEZJGDBE-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1SC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results