accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOOK

CAS RN 60096-23-3

IUPAC Name: potassium 4-(1H-indol-3-yl)butanoate
Synonyms: potassium indolebutyrate
Molecular Weight (g/mol): 241.33
Molecular Formula: C12H12KNO2
InChi Key: KTWDHJYSJOSTSJ-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CCCC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results