accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOHBr

CAS RN 601-84-3

IUPAC Name: 2,6-dibromobenzoic acid
Synonyms: benzoic acid, 2,6-dibromo-
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: HQLOEBRPCVIFCT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results