CAS RN 60124-86-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ PCP-D5 (Phencyclidine-D5) Solution, 100 μg/mL in methanol, Sold by MilliporeSigma™ Supelco™
Phencyclidine-D5 is a stable-labeled internal standard suitable for quantitation of PCP levels in urine, blood or hair by LC/MS or GC/MS for testing applications in urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.