accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFOSOOFFF

CAS RN 60129-85-3

IUPAC Name: 2,3,4,5,6-pentafluorophenyl trifluoromethanesulfonate
Synonyms: 2,3,4,5,6-pentafluorophenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 316.12
Molecular Formula: C7F8O3S
InChi Key: OTTBRWDUQHDNBY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OS(=O)(=O)C(F)(F)F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results