accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60142-89-4

IUPAC Name: 7-{[(tert-butoxy)carbonyl]amino}heptanoic acid
Synonyms: 7-[(tert-butoxycarbonyl)amino]heptanoic acid
Molecular Weight (g/mol): 245.32
Molecular Formula: C12H23NO4
InChi Key: HJENAZQPOGVAEK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results