CAS RN 6015-57-2
IUPAC Name:
N-{4-[(2E)-2,3-dihydro-1,3-benzoxazol-2-ylidene]-3-oxocyclohexa-1,5-dien-1-yl}-3,4,5-triethoxybenzamide
Synonyms:
N-{4-[(2E)-3H-1,3-benzoxazol-2-ylidene]-3-oxocyclohexa-1,5-dien-1-yl}-3,4,5-triethoxybenzamide
Molecular Weight (g/mol):
462.5
Molecular Formula:
C26H26N2O6
InChi Key:
HDSCRNRNYYEKME-NLRVBDNBSA-N
SMILES:
CCOC1=CC(=CC(OCC)=C1OCC)C(=O)NC1=CC(=O)\C(C=C1)=C1/NC2=CC=CC=C2O1
1
–
1
of
1
results
1
Promotions Available
4-Chloro-5-nitrophthalimide, 95%, Thermo Scientific™
4-Chloro-5-nitrophthalimide, 95%, Thermo Scientific™
CAS: 6015-57-2 Molecular Formula: C8H3ClN2O4 Molecular Weight (g/mol): 226.572 MDL Number: MFCD00052331 InChI Key: ADLVDYMTBOSDFE-UHFFFAOYSA-N PubChem CID: 2801129 IUPAC Name: 5-chloro-6-nitroisoindole-1,3-dione SMILES: C1=C2C(=CC(=C1[N+](=O)[O-])Cl)C(=O)NC2=O