accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60166-83-8

IUPAC Name: 3-methoxythiophene-2-carboxylic acid
Synonyms: 3-methoxythiophene-2-carboxylic acid
Molecular Weight (g/mol): 158.17
Molecular Formula: C6H6O3S
InChi Key: LSSMRBBQCHSDNS-UHFFFAOYSA-N
SMILES: COC1=C(SC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results