accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60166-86-1

IUPAC Name: 5-methanesulfonylthiophene-2-carboxylic acid
Synonyms: 5-methanesulfonylthiophene-2-carboxylic acid
Molecular Weight (g/mol): 206.23
Molecular Formula: C6H6O4S2
InChi Key: SLWRTVINHWIGTK-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(S1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results