CAS RN 60177-39-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
344.95
Molecular Formula:
C22H31ClN
InChi Key:
—
SMILES:
[Cl-].*-C=C.*-C=C.*-C=C.C1=CC=CC=C1.C[N+](C)(C)CC1=CC=CC=C1
1
–
2
of
2
results
1
Ion exchanger Dowex™ 1-X8, Cl- form, EMD-Millipore
CAS: 60177-39-1 Molecular Formula: C22H31ClN Molecular Weight (g/mol): 344.95
2
MilliporeSigma™ Supelco™ AmberLite HPR4800 Cl, Chloride Form
Formerly Dowex™ Marathon™ A, chloride form