accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONOO

CAS RN 602-01-7

IUPAC Name: 1-methyl-2,3-dinitrobenzene
Synonyms: 2,3-dinitrotoluene
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: DYSXLQBUUOPLBB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results