accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)O(R)HO(S)HO(S)OH(R)OHNHH3COOOH3CSCH3

CAS RN 602-41-5

IUPAC Name: N-[3,4-dimethoxy-14-(methylsulfanyl)-13-oxo-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
Synonyms: thiocolchicoside
Molecular Weight (g/mol): 563.62
Molecular Formula: C27H33NO10S
InChi Key: LEQAKWQJCITZNK-MSQQGMGVSA-N
SMILES: COC1=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C=C2CCC(NC(C)=O)C3=CC(=O)C(SC)=CC=C3C2=C1OC
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results