accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNNOClCl

CAS RN 60207-90-1

IUPAC Name: 1-{[2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolan-2-yl]methyl}-1H-1,2,4-triazole
Synonyms: propiconazole radar desmel orbit banner caswell no. 323EE epa pesticide chemical code 122101 Tilt
Molecular Weight (g/mol): 342.22
Molecular Formula: C15H17Cl2N3O2
InChi Key: STJLVHWMYQXCPB-UHFFFAOYNA-N
SMILES: CCCC1COC(CN2C=NC=N2)(O1)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results