accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60211-57-6

IUPAC Name: (3,5-dichlorophenyl)methanol
Synonyms: benzenemethanol, 3,5-dichloro-
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: VSNNLLQKDRCKCB-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (4)