accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6025-60-1

IUPAC Name: 2-(1H-pyrrol-1-yl)aniline
Synonyms: 2-(pyrrol-1-yl)aniline
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: GDMZHPUPLWQIBD-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)