accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHN

CAS RN 60298-18-2

IUPAC Name: ethyl 5-oxopyrrolidine-3-carboxylate
Synonyms: ethyl 5-oxopyrrolidine-3-carboxylate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: QWMPXTNXXREMCK-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CNC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results