accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-00-9

IUPAC Name: 7-(2-hydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: proxyphylline purophyllin hydroxypropyltheophylline β-oxypropyltheophyllin monophyllin proxiphylline proxy-retardoral sigophyl 2-hydroxypropyl-7-theophylline proxyphyllinum
Molecular Weight (g/mol): 238.25
Molecular Formula: C10H14N4O3
InChi Key: KYHQZNGJUGFTGR-UHFFFAOYNA-N
SMILES: CC(O)CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results