accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-33-8

IUPAC Name: triphenylbismuthane
Synonyms: bismuthine, triphenyl-
Molecular Weight (g/mol): 440.3
Molecular Formula: C18H15Bi
InChi Key: ZHXAZZQXWJJBHA-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Bi](C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results