accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-45-2

IUPAC Name: 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Synonyms: aurin p-rosolic acid corallin spirit soluble 4,4'-dihydroxyfuchsone aurine rosolic acid aurin no. 555 corallin spirit aurine
Molecular Weight (g/mol): 290.32
Molecular Formula: C19H14O3
InChi Key: FYEHYMARPSSOBO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (4)