accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OH3CO

CAS RN 603-69-0

IUPAC Name: ethyl 2-acetyl-3-oxobutanoate
Synonyms: ethyl 2-acetyl-3-oxobutanoate
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: YMCDYRGMTRCAPZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(C)=O)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)