accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-76-9

IUPAC Name: 1-methyl-1H-indole
Synonyms: 1-methylindole
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: BLRHMMGNCXNXJL-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC=C12
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)