accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-85-0

IUPAC Name: 2-amino-3-nitrophenol
Synonyms: phenol, 2-amino-3-nitro-
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: KUCWUAFNGCMZDB-UHFFFAOYSA-N
SMILES: NC1=C(O)C=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)