CAS RN 603108-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Glucokinase Activator, Cpd A
The Glucokinase Activator, Cpd A, also referenced under CAS 603108-44-7, modulates Glucokinase. This small molecule/inhibitor is primarily used for Activators/Inducers applications.