accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3BOHHO

CAS RN 603122-41-4

IUPAC Name: [3-methoxy-4-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 3-methoxy-4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 209.99
Molecular Formula: C9H11BO5
InChi Key: YCXPWNGIPLGCOJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1OC)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results