CAS RN 603122-80-1
IUPAC Name:
[2-chloro-4-(methoxycarbonyl)phenyl]boronic acid
Synonyms:
2-chloro-4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol):
214.41
Molecular Formula:
C8H8BClO4
InChi Key:
ITEQHBSWRMYSRP-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=C(C=C1)B(O)O