accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603122-84-5

IUPAC Name: [2-fluoro-4-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 2-fluoro-4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 197.96
Molecular Formula: C8H8BFO4
InChi Key: BKWRLCIYMAYFPA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results