accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSOOFFFOCH3

CAS RN 60319-09-7

IUPAC Name: 3,5-dimethoxyphenyl trifluoromethanesulfonate
Synonyms: 3,5-dimethoxyphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 286.22
Molecular Formula: C9H9F3O5S
InChi Key: XCOBIQQRDHVTAN-UHFFFAOYSA-N
SMILES: COC1=CC(OS(=O)(=O)C(F)(F)F)=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results