accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOCH3

CAS RN 6034-46-4

IUPAC Name: 2-(acetyloxy)propanoic acid
Synonyms: 2-(acetyloxy)propanoic acid
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: WTLNOANVTIKPEE-UHFFFAOYNA-N
SMILES: CC(OC(C)=O)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results