accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6035-47-8

H2OH2ONaHOSOO

CAS RN 6035-47-8

IUPAC Name: sodium dihydrate sulfinomethanolate
Synonyms: sodium dihydrate sulfinomethanolate
Molecular Weight (g/mol): 154.11
Molecular Formula: CH7NaO5S
InChi Key: VJWVWUHJLZEHOU-UHFFFAOYNA-N
SMILES: O.O.[Na+].OS(=O)C[O-]
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)