accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60459-08-7

IUPAC Name: λ²-cobalt(2+) sulfate
Synonyms: cobalt sulfate|cobalt(II) sulfate|cobaltous sulfate|CoSO4|cobalt brown|cobalt (2+) sulfate
Molecular Weight (g/mol): 154.99
Molecular Formula: CoO4S
InChi Key: KTVIXTQDYHMGHF-UHFFFAOYSA-L
SMILES: [Co++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)