CAS RN 60469-70-7
IUPAC Name:
hydrogen 2-methylpropan-2-amine bromide
Synonyms:
hydrogen erbumine bromide
Molecular Weight (g/mol):
154.05
Molecular Formula:
C4H12BrN
InChi Key:
CQKAPARXKPTKBK-UHFFFAOYSA-N
SMILES:
[H+].[Br-].CC(C)(C)N
1
–
1
of
1
results
1
tert-Butylamine Hydrobromide 98.0+%, TCI America™
CAS: 60469-70-7 Molecular Formula: C4H12BrN Molecular Weight (g/mol): 154.05 MDL Number: MFCD00674188 InChI Key: CQKAPARXKPTKBK-UHFFFAOYSA-N Synonym: tert-Butylammonium Bromide PubChem CID: 3085186 IUPAC Name: hydrogen 2-methylpropan-2-amine bromide SMILES: [H+].[Br-].CC(C)(C)N