CAS RN 60477-38-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Pifithrin-α, p-Nitro, Cyclic
A cell-permeable p53 inhibitor that exhibits 10-fold higher potency and 50% longer half-life than Pifithrin-α.