accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6048-29-9

IUPAC Name: (2-oxo-2-phenylethyl)triphenylphosphanium bromide
Synonyms: (2-oxo-2-phenylethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 461.34
Molecular Formula: C26H22BrOP
InChi Key: AEHDSYHVTDJGDN-UHFFFAOYSA-M
SMILES: [Br-].O=C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results