accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60484-67-5

IUPAC Name: 4-heptylbenzonitrile
Synonyms: 4-heptylbenzonitrile
Molecular Weight (g/mol): 201.31
Molecular Formula: C14H19N
InChi Key: XTIKBCXMOYZUMG-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results