accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 605-39-0

IUPAC Name: 2,2'-dimethyl-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 2,2'-dimethyl-
Molecular Weight (g/mol): 182.27
Molecular Formula: C14H14
InChi Key: ABMKWMASVFVTMD-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results