accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOOH3CCH3

CAS RN 605-45-8

IUPAC Name: 1,2-bis(propan-2-yl) benzene-1,2-dicarboxylate
Synonyms: diisopropyl phthalate di-I-propyl phthalate
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: QWDBCIAVABMJPP-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=CC=CC=C1C(=O)OC(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)