accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 605-48-1

IUPAC Name: 9,10-dichloroanthracene
Synonyms: 9,10-dichloroanthracene
Molecular Weight (g/mol): 247.12
Molecular Formula: C14H8Cl2
InChi Key: FKDIWXZNKAZCBY-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=C(Cl)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results