accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 605-61-8

IUPAC Name: 1-chloro-4-nitronaphthalene
Synonyms: 1-chloro-4-nitronaphthalene
Molecular Weight (g/mol): 207.61
Molecular Formula: C10H6ClNO2
InChi Key: QLIOCENRPBJEPI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CC=CC2=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results