accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOO

CAS RN 605-62-9

IUPAC Name: 4-nitronaphthalen-1-ol
Synonyms: 4-nitronaphthalen-1-ol
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: AUIRNGLMBHIITH-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results