accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 605-65-2

IUPAC Name: 5-(dimethylamino)naphthalene-1-sulfonyl chloride
Synonyms: dansyl|dns chloride
Molecular Weight (g/mol): 269.74
Molecular Formula: C12H12ClNO2S
InChi Key: XPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
Quantity 
  • (4)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Color 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (4)
  • (6)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)