accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOO

CAS RN 605-70-9

IUPAC Name: naphthalene-1,4-dicarboxylic acid
Synonyms: naphthalene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 216.19
Molecular Formula: C12H8O4
InChi Key: ABMFBCRYHDZLRD-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)