accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6051-43-0

IUPAC Name: 4-carbamoylbenzoic acid
Synonyms: 4-carbamoylbenzoic acid
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: JMHSCWJIDIKGNZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results