CAS RN 60548-09-6
IUPAC Name:
1-(furan-2-carbonyl)piperazine hydrochloride
Synonyms:
1-(furan-2-carbonyl)piperazine hydrochloride
Molecular Weight (g/mol):
216.67
Molecular Formula:
C9H13ClN2O2
InChi Key:
FMFUHCXDFVDINI-UHFFFAOYSA-N
SMILES:
Cl.O=C(N1CCNCC1)C1=CC=CO1
1
–
1
of
1
results
1
1-(2-Furoyl)piperazine Hydrochloride 98.0+%, TCI America™
CAS: 60548-09-6 Molecular Formula: C9H13ClN2O2 Molecular Weight (g/mol): 216.67 MDL Number: MFCD00236012 InChI Key: FMFUHCXDFVDINI-UHFFFAOYSA-N Synonym: 1-[(2-Furan)carbonyl]piperazine Hydrochloride PubChem CID: 12289910 IUPAC Name: 1-(furan-2-carbonyl)piperazine hydrochloride SMILES: Cl.O=C(N1CCNCC1)C1=CC=CO1