accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6055-52-3

IUPAC Name: hexane-1,6-bis(aminium) dichloride
Synonyms: hexane-1,6-bis(aminium) dichloride
Molecular Weight (g/mol): 189.12
Molecular Formula: C6H18Cl2N2
InChi Key: XMVQMBLTFKAIOX-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[NH3+]CCCCCC[NH3+]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)